Learn More
Greiner Bio-One MiniCollect™ Tube, Red Cap, CAT Serum Clot Activator, Non-Sterile
Product Code. 15788169
Change view
Click to view available options
Quantity:
500/Pk.
Unit Size:
Pack of 500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15788169 500/Pk. Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15788169 Supplier Greiner Bio-One Supplier No. 450534
  • ONLINE EXCLUSIVE PRICE
    987.00€
    861.49€ / 500 pièces
    Économisez 125.51€
    12% de réduction
 Restricted Item
Expédition estimée: 09-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Includes inner wall coated with microscopic silica particles to activate coagulation.
  • The inner wall of the serum tube is specially coated with microscopic silica particles, which activate the coagulation process.
TRUSTED_SUSTAINABILITY
Specifications
Color Red
Additive CAT Serum Sep Clot Activator
Sterility Non-sterile
Capacity (Metric) 0.5 mL, 1.0 mL
Closure Included Yes
Closure Color Red
Quantity 500/Pk.