Learn More
Endecotts™ Minor 200 Sieve Shaker
Product Code. 15359001
Change view
Click to view available options
Particle Size:
38 μm to 125 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Particle Size Unit Size
15359001 38 μm to 125 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 15359001 Supplier Endecotts™ Supplier No. MIN200/23050
  • ONLINE EXCLUSIVE PRICE
    2285.00€
    1790.85€ / 1 pièces
    Économisez 494.15€
    21% de réduction
View all promotions
Expédition estimée: 14-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Compact and portable. Endecotts Minor 200 Sieve Shaker is ideal for use in laboratories and plants.
  • Compact and portable and easy to use
  • Electromagnetic drive for quiet and virtually maintenance free operation
  • Requires only small storage space due to small footprint and easily removable clamping belt system (included)
  • Different voltages available
Compliance
AASHTO T 27
Specifications
Electrical Requirements 230 V, 50 Hz
Product Type Sieve Shaker
Diameter (Metric) 100/200 mm
Model Minor 200 Benchtop
No. of Sieves 8 full height/16 half height (200 mm sieves)
Particle Size 38 μm to 125 mm
Timer Range Analog, 0 to 60 min., Continuous