Learn More
Edulab Mirror, Convex - Pack of 10
Product Code. 30280303
Change view
Click to view available options
Product Type:
Convex Mirror
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
30280303 Convex Mirror Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30280303 Supplier Edulab Supplier No. PH0526D
  • ONLINE EXCLUSIVE PRICE
    17.80€
    17.26€ / 10 pièces
    Économisez 0.54€
    3% de réduction
Expédition estimée: 09-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Convex mirror designed for classroom optics experiments and practical demonstrations.
  • 50 mm diameter convex mirror for practical reflection experiments
  • Optically worked to provide clear and accurate reflections
  • Focal length of 150 mm for standard classroom use
  • Silvered back with protective coating for durability
Specifications
Product Type Convex Mirror