Learn More
Emma Safety Footwear Mitchel XL Safety Shoes
Product Code. 15988201
Change view
Click to view available options
Size:
52
53
Unit Size:
1 pair
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15988201 53 1 pair
15978201 52 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15988201 Supplier Emma Safety Footwear Supplier No. MM13035853
  • ONLINE EXCLUSIVE PRICE
    299.69€
    177.45€ / 1 paires
    Économisez 122.24€
    40% de réduction
Expédition estimée: 16-03-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Ideal for working with metal with optimal comfort, ventilation and moisture removal technology. A perfectly integrated steel toe protects feet against falling pieces of metal and other exterior threats. The Gripforce™ DUO soles withstand high temperatures and are oil and petrol resistant.
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • Nitrile Rubber Heat Resistant Outsole
Specifications
Certifications/Compliance EN ISO 20345:2011, EN 12568:2010
Closure Type Laces
Color Black
Design Mitchel XL
Liner Material Full grain Nappa leather / Split leather
Material Leather
Size 53
Toe Material Steel