Learn More
Mitomycin A, TRC
Product Code. 30007923
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.50mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007923 2.5 mg 2.50mg
30004582 10 mg 10mg
30009060 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007923 Supplier Toronto Research Chemicals Supplier No. TRCM3718902.5MG
 Restricted Item
Expédition estimée: 09-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4055-39-4
Molecular Formula C16 H19 N3 O6
Molecular Weight (g/mol) 349.34
Synonym Azirino[2',3':3,4]pyrrolo[1,2-a]indole-4,7-dione, 8-[[(aminocarbonyl)oxy]methyl]-1,1a,2,8,8a,8b-hexahydro-6,8a-dimethoxy-5-methyl-, (1aS,8S,8aR,8bS)-, Azirino[2',3':3,4]pyrrolo[1,2-a]indole-4,7-dione, 1,1a,2,8,8a,8b-hexahydro-8-(hydroxymethyl)-6,8a-dimethoxy-5-methyl-, carbamate (ester) (8CI), Azirino[2',3':3,4]pyrrolo[1,2-a]indole-4,7-dione, 8-[[(aminocarbonyl)oxy]methyl]-1,1a,2,8,8a,8b-hexahydro-6,8a-dimethoxy-5-methyl-, [1aS-(1aα,8β,8aα,8bα)]-, Mitomycin A (7CI), (1aS,8S,8aR,8bS)-8-[[(Aminocarbonyl)oxy]methyl]-1,1a,2,8,8a,8b-hexahydro-6,8a-dimethoxy-5-methylazirino[2',3':3,4]pyrrolo[1,2-a]indole-4,7-dione, NSC 75986, Mitomycin Imp. B (EP)
SMILES COC1=C(C)C(=O)C2=C([C@@H](COC(=O)N)[C@@]3(OC)[C@H]4N[C@H]4CN23)C1=O
Specifications
Quantity 2.5 mg
InChI Formula InChI=1S/C16H19N3O6/c1-6-11(20)10-9(12(21)13(6)23-2)7(5-25-15(17)22)16(24-3)14-8(18-14)4-19(10)16/h7-8,14,18H,4-5H2,1-3H3,(H2,17,22)/t7-,8+,14+,16-/m1/s1
Formula Weight 349.1274
Percent Purity >95%
Chemical Name or Material Mitomycin A
Grade HPLC