Learn More
SP Genevac™ miVac™ Replacement lid seal
Product Code. 18897304
Change view
Click to view available options
For Use With (Equipment):
Duo Concentrator
Quattro Concentrator
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
18897304 Duo Concentrator Each
31723036 Quattro Concentrator Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18897304 Supplier SP Genevac™ Supplier No. 600001
 Restricted Item
Expédition estimée: 28-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Replacement Seal for miVac Concentrator
  • Designed as a direct replacement lid seal for miVac concentrators to maintain optimal sealing performance
  • Compatible with both miVac Duo and Quattro Concentrator systems for versatile use
  • Helps ensure proper vacuum conditions for efficient sample concentration
  • Manufactured for reliable fit and long-lasting performance in laboratory environments
Specifications
Material EPDM
Type Seal
For Use With (Equipment) Duo Concentrator