Learn More
R&D Systems™ ML 281
Product Code. 30002066
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30002066 10 mg 10mg
30001984 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30002066 Supplier R&D Systems™ Supplier No. 4880/10
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
A Tocris product, part of R&D Systems. STK33 inhibitor
Inhibitor of the serine/threonine kinase STK33 (IC50 = 14 nM). Exhibits >700-fold selectivity for STK33 over the structurally related protein kinase A (PKA); also exhibits 550-fold selectivity over Aurora kinase B.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1404437-62-2
Molecular Formula C22H19N3O2S
Specifications
Quantity 10 mg
Product Line Tocris
Formula Weight 389.47
Purity >98%
Molecular Weight (g/mol) 389.473
Chemical Name or Material ML 281