Learn More
SymLine Module Signal Set (EU)
Product Code. 17347419
Change view
Click to view available options
For Use With (Application):
Safety Cabinets
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
17347419 Safety Cabinets Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 17347419 Supplier SymLine Supplier No. 106678
  • ONLINE EXCLUSIVE PRICE
    722.00€
    618.28€ / 1 pièces
    Économisez 103.72€
    14% de réduction
Expédition estimée: 04-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Monitor two filling levels at once, perfect integration into lab furniture with the Switch Amplifier
  • Connect NAMUR-Sensors or potential-free mechanical contacts which are mounted in the zones 0, 1, 2 (Gas) or 20, 21, 22 (dust)
  • Compact design
Specifications
Description Module Signalset for safety cabinets 'EU-plug', consisting of: 1 x Switch amplifier EU, 2 channels (108278) 1 x Housing for the isolating switch amplifier (106590) 2 x Cable set: Connection for switch amplifier - Built-in signal box (108314)
For Use With (Application) Safety Cabinets
Product Type Module Signal Set (EU)
Includes 1 x Switch amplifier EU, 2 channels (108278) 1 x Housing for the isolating switch amplifier (106590) 2 x Cable set: Connection for switch amplifier - Built-in signal box (108314)