Learn More
Invitrogen™ CultureWell™ Chambered Coverglass for Cell Culture, 16 wells per coverglass
Product Code. 10474022
Change view
Click to view available options
Quantity:
1 set
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10474022 1 set Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10474022 Supplier Invitrogen™ Supplier No. C37000
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Ideal for cell culture and fluorescence imaging applications
The CultureWell removable chambered coverglass provided sterile and ready to use—contains 16 wells that can each hold up to 250μL, allowing cells to be cultured in a number of different conditions on a single slide. When the cells are ready to be imaged, coverglass removal is made easy by the use of a simple tool (Cat. No. C37010, purchased separately) that separates the parts without the need for excessive force, eliminating the risk of coverglass breakage. Frosted microscope slides are also provided for mounting. A sample size containing two chambered coverglasses and the removal tool is also available (Cat. No. C37005)
Cell Analysis, Cell Culture, Cellular Imaging, IHC Sample Preparation, IHC Staining & Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Sample Preparation, Immunohistochemistry (IHC), Mammalian Cell Culture
TRUSTED_SUSTAINABILITY
Specifications
Type Coverglass
Wells 16
Culture Area 0.4 cm2
Surface Treatment Cell Culture
Volume (Metric) Working 250 μL
Content And Storage Store at room temperature.
Shipping Condition Room Temperature
Sterility Sterile
Quantity 1 set
For Research Use Only. Not for use in diagnostic procedures.