Learn More
Invitrogen™ Dihydroethidium (Hydroethidine)
Product Code. 10174952
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10174952 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10174952 Supplier Invitrogen™ Supplier No. D23107
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
En stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy
Free radical sensor
The superoxide indicator dihydroethdium, also called hydroethidine, exhibits blue-fluorescence in the cytosol until oxidized, where it intercalates within the cell's DNA, staining its nucleus a bright fluorescent red.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 mL
For Use With (Application) Superoxide Indicator
No. of Tests 1 mL
Detection Method Fluorescence
Excitation/Emission 518/606 nm
Product Type ROS Indicator
Excitation Wavelength Range 518 nm
Emission Wavelength Range 606 nm
For Research Use Only. Not for use in diagnostic procedures.