Learn More
Mouse skin tissue lysate - total protein (0 days), 100 ug
Product Code. 32155297 Shop All Abcam Products
Change view
Click to view available options
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
32155297 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32155297 Supplier Abcam Supplier No. ab7269100ug
 Restricted Item
Expédition estimée: 07-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Mouse skin cell/tissue lysate.
  • Species: Mouse
  • Tissue: skin
  • Form: Liquid
Specifications
For Use With (Application) Western Blot