Learn More
MP Biomedicals™ γ-Globulin, Human Plasma
Product Code. 11414276 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11414276 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11414276 Supplier MP Biomedicals™ Supplier No. 08823102
  • ONLINE EXCLUSIVE PRICE
    418.24€
    382.20€ / 10g
    Économisez 36.04€
    8% de réduction
 Restricted Item
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Each donor unit used in the preparation of the product was tested and found negative for Anti-HIV 1/2, HBsAg, Anti-HCV and HIV-1 Ag by FDA approved methods or CE approved methods.
Specifications
Color White
pH 6.44
Quantity 10 g
Content And Storage 2°C to 8°C
Form Powder
For Use With (Application) Used as an immunological challenge in studies of mechanisms of immunotolerance
Purity or Quality Grade >96%
Product Type Human Plasma gamma Globulin