Learn More
MP Biomedicals™ Hand-Pump for 1-GAL 7X™ Bottles
Product Code. 15749180 Shop All MP Biomedicals Products
Change view
Click to view available options
Volume (English):
1 gal.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (English) Unit Size
15749180 1 gal. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15749180 Supplier MP Biomedicals™ Supplier No. 0976680HP
  • ONLINE EXCLUSIVE PRICE
    12.80€
    11.55€ / 1 pièces
    Économisez 1.25€
    9% de réduction
View all promotions
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Accessory for 7X™ cleaning solution
White hand pump.
  • Hand pump for 1 gallon 7X™ bottles
Specifications
Appearance White
Volume (English) 1 gal.