Learn More
FTM Technologies™ Multi-Operator Fume Hoods
Product Code. 11778044
Change view
Click to view available options
Includes:
4 extraction inputs
6 extraction inputs
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
11778044 4 extraction inputs Each
11778054 6 extraction inputs Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 11778044 Supplier FTM Technologies™ Supplier No. HI4P4S
  • ONLINE EXCLUSIVE PRICE
    1510.00€
    1255.22€ / 1 pièces
    Économisez 254.78€
    16% de réduction
 Restricted Item
Expédition estimée: 07-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Used for filtration where extraction arms or other accessories are needed. FTM Technologies™ Multi-Operator Fume Hoods provide multi-workstation filtration for welding fumes, solvents and other hazardous substances.
  • Available with either 4 or 6 extraction inputs
  • Delivered with ringed PVC pipe
  • Compatible with the use of prefilters
  • Can be used as a recirculating (ductless) fume hood, using active carbon filtration or dust filter, or with an external output (external filter plus exhaust duct)
Specifications
Dimensions (L x W x H) 350 x 200 x 430 mm
Flow Rate 50 to 450 m3/h
Includes 4 extraction inputs
Voltage 230 V
Weight (Metric) 6 kg
Frequency 50 Hz