Learn More
WTW™ Multi-Parameter Meters
Product Code. 15205699
Change view
Click to view available options
Measurement Modes:
pH, ORP, Conductivity, Dissolved Oxygen
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Modes Unit Size
15205699 pH, ORP, Conductivity, Dissolved Oxygen Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 15205699 Supplier WTW™ Supplier No. 2FA310
  • ONLINE EXCLUSIVE PRICE
    1985.00€
    1717.00€ / 1 pièces
    Économisez 268.00€
    13% de réduction
View all promotions
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Ideal for measuring environmental applications in ground and surface water measurement, aquaculture, and waste water treatment plants.
  • Automatic/Manual calibration for Multi-Parameter Meter Model Multi 3320
  • Includes IP 67 Waterproof, Mini USB-B Interface, 4 x 1.5 V AA Power supply
  • For use with measurement of pH, ORP, conductivity, and dissolved oxygen (electro chemical.)
  • Environmental applications in ground and surface water measurement, aquaculture, waste water treatment plants
TRUSTED_SUSTAINABILITY
Specifications
Accuracy (ISE) 0.000 to 999999μmol/L
Battery Type AA
Calibration Automatic/Manual
Calibration Points (pH) ±2.000 to19,999
Display Type LCD Graphic, Backlit
IP Rating IP67
Measurement Modes pH, ORP, Conductivity, Dissolved Oxygen
Range (Conductivity) 0.000 to 1000 mS/cm
Range (mV) -2500 to 2500 mV
Range (pH) -2.000 to 19.999
Range (Salinity) 0.0 to 70.0
Range (TDS) 0 to 199.9 g/L
For Use With (Application) Measurement of pH, ORP, conductivity, and dissolved oxygen (electro chemical.) Environmental applications in ground and surface water measurement, aquaculture, waste water treatment plants.
Includes IP 67 Waterproof, Mini USB-B Interface, 4 x 1.5 V AA Power supply
Inputs Two
Memory Up to 10 calibration records
Model Multi 3320
Power Supply 1.5V
Product Type Multi-Parameter Meter
Range (ISE) 0.000 to 999999μmol/l
TDS Factor Range 0 to 199.9g/L