Learn More
WTW™ MultiLab™ Pro 40 IDS WL
Product Code. 30929043
Change view
Click to view available options
Channels:
4
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Channels Unit Size
30929043 4 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 30929043 Supplier WTW™ Supplier No. 1FD78W
 Restricted Item
Expédition estimée: 21-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Xylem’s MultiLab™ Pro IDS is a modern high-performance multiparameter device with 4 galvanically isolated measuring channels for pH, ORP, mV, conductivity, oxygen, ISE and turbidity measurement.
The MultiLab™ Pro IDS from Xylem is a modern multi-parameter measuring device with up to four measurement channels.
  • Ease of use
  • LIMS ready
  • GxP compliance
  • Compact and functional design
Highlights:
  • Measures pH, ORP, conductivity, dissolved oxygen, ISE (coming soon) and turbidity
  • Wireless-ready for IDS modules
  • Digital sensor recognition
  • Simultaneous measurement with four independent measuring channels
  • Brilliant touch display
  • Preset and definable buffer sets
  • QSC calibration
  • AutoRead function
  • Export to PDF/A, JSON and CSV
  • Special touchscreen glass can be operated with gloves and resistant to alcohol and disinfectants
Scope of delivery:
  • Device with stand and power supply, IDS WLM-Kit (wireless modules)
TRUSTED_SUSTAINABILITY
Specifications
Calibration pH
Certifications/Compliance CE Certified
Channels 4
Display Type Color TFT
IP Rating IP42
Measurement Modes pH, Redox, mV, Conductivity, Oxygen
Range (Conductivity) 0.001 μS/cm to 200 μS/cm
Range (pH) 0.000 to 14.000
Resolution (ISE) depending on Sensor
Resolution (pH) depending on Sensor
Temperature Range (Metric) depending on Sensor
Log Function Type Manual or Interval
Product Type Multiparameter Laboratory Measuring Device
Voltage 115/230 VAC