Learn More
Melford Mycophenolic Acid
Product Code. 30018547
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018547 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30018547 Supplier Melford Supplier No. M930000.1
 Restricted Item
Expédition estimée: 11-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Immunosuppressive agent.
Immunosuppressive agent. Suppresses cylokine induced nitric oxide production. Used to select animal cells expressing the E. Coli. gene for xanthine guanine phosphoribosyl transferase.
Specifications
CAS 24280-93-1
Molecular Formula C17H20O6
Quantity 100 mg