Learn More
N-(1-Cyano-1-methylethyl)-2-methoxyethanesulfonamide, TRC
Product Code. 30351391
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30351391 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30351391 Supplier Toronto Research Chemicals Supplier No. TRCC212050100MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-(1-Cyano-1-methylethyl)-2-methoxyethanesulfonamide
CAS 103472-22-6
Molecular Formula C7 H14 N2 O3 S
Quantity 100 mg
Synonym N-(2-cyanopropan-2-yl)-2-methoxyethane-1-sulfonamide
InChI Formula InChI=1S/C7H14N2O3S/c1-7(2,6-8)9-13(10,11)5-4-12-3/h9H,4-5H2,1-3H3
SMILES COCCS(=O)(=O)NC(C)(C)C#N
IUPAC Name N-(2-cyanopropan-2-yl)-2-methoxyethanesulfonamide
Molecular Weight (g/mol) 206.263
Formula Weight 206.073