missing translation for 'onlineSavingsMsg'
Learn More

N-(1-Naphthyl)-4-nitrobenzenesulfonamide, 97%, Thermo Scientific™

Product Code. 15424398
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15424398 100 mg 100mg
15434398 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15424398 Supplier Thermo Scientific Alfa Aesar Supplier No. H58020.MC

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56799-93-0
Molecular Formula C16H12N2O4S
Molecular Weight (g/mol) 328.342
MDL Number MFCD01985189
InChI Key PQKIMVCBRAJXGP-UHFFFAOYSA-N
Synonym n-naphthalen-1-yl-4-nitrobenzenesulfonamide, n-1-naphthyl-4-nitrobenzenesulfonamide, cbmicro_011238
PubChem CID 879693
IUPAC Name N-naphthalen-1-yl-4-nitrobenzenesulfonamide
SMILES C1=CC=C2C(=C1)C=CC=C2NS(=O)(=O)C3=CC=C(C=C3)[N+](=O)[O-]

Specifications

Quantity 100 mg
Beilstein 3003355
Formula Weight 328.34
Percent Purity 97%
Chemical Name or Material N-(1-Naphthyl)-4-nitrobenzenesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.