missing translation for 'onlineSavingsMsg'
Learn More

N-(2,4-Difluorophenyl)maleamic acid, 97%, Thermo Scientific™

Product Code. 11456616
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11456616 5 g 5g
11446616 1 g 1g
11466616 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11456616 Supplier Thermo Scientific Alfa Aesar Supplier No. B24837.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6954-64-9
Molecular Formula C10H7F2NO3
Molecular Weight (g/mol) 227.167
MDL Number MFCD00082911
InChI Key DVBSHLGHHLTWPZ-ARJAWSKDSA-N
Synonym n-2,4-difluorophenyl maleamic acid, 2z-3-2,4-difluorophenyl carbamoyl prop-2-enoic acid, 2-butenoic acid,4-2,4-difluorophenyl amino-4-oxo-, 2z, n-2,4-difluorophenyl maleamic acid, n-2,4-difluorphenyl-maleamsaeure, z-4-2,4-difluoroanilino-4-oxo-2-butenoic acid, z-4-2,4-difluoroanilino-4-oxobut-2-enoic acid, z-4-2,4-difluorophenyl amino-4-oxobut-2-enoic acid, 2z-3-n-2,4-difluorophenyl carbamoyl prop-2-enoic acid, z-4-2,4-bis fluoranyl phenyl amino-4-oxidanylidene-but-2-enoic acid
PubChem CID 1715171
IUPAC Name (Z)-4-(2,4-difluoroanilino)-4-oxobut-2-enoic acid
SMILES C1=CC(=C(C=C1F)F)NC(=O)C=CC(=O)O

Specifications

Melting Point 190°C to 192°C
Quantity 5 g
Formula Weight 227.16
Percent Purity 97%
Chemical Name or Material N-(2,4-Difluorophenyl)maleamic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.