Learn More
N-(2,6-Dimethylphenyl)-2-(2-oxopyrrolidin-1-yl)acetamide, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 77191-36-7 |
| Molecular Formula | C14H18N2O2 |
| MDL Number | MFCD00209882 |
| Quantity | 250 mg |
| SMILES | O=C(NC1=C(C)C=CC=C1C)CN2C(CCC2)=O |
| Molecular Weight (g/mol) | 246.30 |
| Percent Purity | 97% |
| Chemical Name or Material | N-(2,6-Dimethylphenyl)-2-(2-oxopyrrolidin-1-yl)acetamide |