Learn More
N-[[4-[[[4-[[tert-Butyloxycarbonyl]amino][1,1'-biphenyl]-3-yl]amino]carbonyl]phenyl]methyl]carbamic Acid Methyl Ester-d3, TRC
Product Code. 30348663
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30348663 100 mg 100mg
30335015 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30348663 Supplier Toronto Research Chemicals Supplier No. TRCB693442100MG
 Restricted Item
Expédition estimée: 09-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-((4-(((4-((tert-Butyloxycarbonyl)amino)(1,1'-biphenyl)-3-yl)amino)carbonyl)phenyl)methyl)carbamic Acid Methyl Ester-d3
Molecular Formula C27H26D3N3O5
Quantity 100 mg
Synonym N-[[4-[[[4-[[(1,1-Dimethylethoxy)carbonyl]amino][1,1'-biphenyl]-3-yl]amino]carbonyl]phenyl]methyl]carbamic Acid Methyl Ester-d3
InChI Formula InChI=1S/C27H29N3O5/c1-27(2,3)35-26(33)30-22-15-14-21(19-8-6-5-7-9-19)16-23(22)29-24(31)20-12-10-18(11-13-20)17-28-25(32)34-4/h5-16H,17H2,1-4H3,(H,28,32)(H,29,31)(H,30,33)/i4D3
SMILES O=C(NC1=CC(C2=CC=CC=C2)=CC=C1NC(OC(C)(C)C)=O)C3=CC=C(CNC(OC([2H])([2H])[2H])=O)C=C3
IUPAC Name methyl-d3 (4-((4-((tert-butoxycarbonyl)amino)-[1,1'-biphenyl]-3-yl)carbamoyl)benzyl)carbamate
Molecular Weight (g/mol) 478.55
Formula Weight 478.23