Learn More
N-[4-Cyano-3-(trifluoromethyl)phenyl]-2-methylacrylamide, TRC
Product Code. 30366408
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30366408 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30366408 Supplier Toronto Research Chemicals Supplier No. TRCC98819325G
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-[4-Cyano-3-(trifluoromethyl)phenyl]-2-methylprop-2-enamide
CAS 90357-53-2
Molecular Formula C12 H9 F3 N2 O
Quantity 25 g
Synonym 2-Propenamide, N-[4-cyano-3-(trifluoromethyl)phenyl]-2-methyl-, N-[4-Cyano-3-(trifluoromethyl)phenyl]-2-methyl-2-propenamide, N-[4-Cyano-3-(trifluoromethyl)phenyl]-2-methylacrylamide, N-[4-Cyano-3-(trifluoromethyl)phenyl]-2-methylprop-2-enamide
InChI Formula InChI=1S/C12H9F3N2O/c1-7(2)11(18)17-9-4-3-8(6-16)10(5-9)12(13,14)15/h3-5H,1H2,2H3,(H,17,18)
SMILES CC(=C)C(=O)Nc1ccc(C#N)c(c1)C(F)(F)F
IUPAC Name N-[4-cyano-3-(trifluoromethyl)phenyl]-2-methylprop-2-enamide
Molecular Weight (g/mol) 254.2079
Formula Weight 254.0667