Learn More
N-(4-Nitro-2-phenoxyphenyl)methanesulfonamide, 98%, BLD Pharm™
Product Code. 31003653
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
10 g
25 g
Unit Size:
100mg
10g
1g
25g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31003653 5 g 5g
30996343 10 g 10g
30995565 1 g 1g
30990991 100 mg 100mg
30999494 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31003653 Supplier BLD Pharm Supplier No. BD146295g
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 51803-78-2
Molecular Formula C13H12N2O5S
MDL Number MFCD00079470
Quantity 5 g
SMILES CS(=O)(NC1=CC=C([N+]([O-])=O)C=C1OC2=CC=CC=C2)=O
Molecular Weight (g/mol) 308.31
Percent Purity 98%
Chemical Name or Material N-(4-Nitro-2-phenoxyphenyl)methanesulfonamide