Learn More
N-Acetyl-DL-methionine, 98%, BLD Pharm™
Product Code. 31032619
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Unit Size:
100g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31032619 1 kg -
31019213 100 g 100g
31016855 25 g -
31014982 500 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31032619 Supplier BLD Pharm Supplier No. BD1429931000g
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 1115-47-5
Molecular Formula C7H13NO3S
MDL Number MFCD00008681
Quantity 1 kg
SMILES [H]C(CCSC)(NC(C)=O)C(O)=O
Molecular Weight (g/mol) 191.25
Percent Purity 98%
Chemical Name or Material N-Acetyl-DL-methionine