Learn More
N-caproicacidsodiumsalt, 99%, BLD Pharm™
Specifications
Specifications
| CAS | 10051-44-2 |
| Molecular Formula | C6H11NaO2 |
| MDL Number | MFCD00059056 |
| Quantity | 25 g |
| SMILES | CCCCCC([O-])=O.[Na+] |
| Molecular Weight (g/mol) | 138.14 |
| Percent Purity | 99% |
| Chemical Name or Material | N-caproicacidsodiumsalt |