Learn More
N-Cyanoacetylurethane-13C3,15N2, TRC
Product Code. 30362479
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30362479 50 mg 50mg
30353007 25 mg 25mg
30358702 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30362479 Supplier Toronto Research Chemicals Supplier No. TRCC95540250MG
 Restricted Item
Expédition estimée: 09-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
Molecular Formula 13C3 C3 H8 15N N O3
Molecular Weight (g/mol) 160.111
Synonym N-(2-Cyanoacetyl)carbamic Acid Ethyl Ester-13C3,15N2, (Cyanoacetyl)carbamic Acid Ethyl Ester-13C3,15N2, Cyanoacetylurethan-13C3,15N2, Ethyl N-(cyanoacetyl)carbamate-13C3,15N2, Ethyl Cyanoacetylcarbamate-13C3,15N2, N-Carbethoxy-2-cyanoacetamide-13C3,15N2, N-Cyanoacetylurethane-13C3,15N2, NSC 59709-13C3,15N2
IUPAC Name ethyl N-(2-cyanoacetyl)carbamate
SMILES CCO[13C](=O)[15NH][13C](=O)[13CH2]C#N
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C6H8N2O3/c1-2-11-6(10)8-5(9)3-4-7/h2-3H2,1H3,(H,8,9,10)/i3+1,5+1,6+1,8+1
Formula Weight 160.061
Chemical Name or Material N-Cyanoacetylurethane-13C3,15N2