missing translation for 'onlineSavingsMsg'
Learn More

N-Glycyl-L-tyrosine, 98%

Product Code. 18891796 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
18891796 5 g
17113076 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18891796 Supplier Thermo Scientific Alfa Aesar Supplier No. H65388.06
  • ONLINE EXCLUSIVE PRICE
    71.55€
    58.65€ / 5g
    Économisez 12.90€
    18% de réduction
 Restricted Item
En stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 658-79-7 | C11H14N2O4 | 238.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 658-79-7
Molecular Formula C11H14N2O4
Molecular Weight (g/mol) 238.24
MDL Number MFCD00008126
InChI Key XBGGUPMXALFZOT-IMWMWJONNA-N
Synonym glycyl-l-tyrosine, gly-tyr, n-glycyl-l-tyrosine, h-gly-tyr-oh, glycyltyrosine, l-tyrosine, glycyl, s-2-2-aminoacetamido-3-4-hydroxyphenyl propanoic acid, chembl53400, 2s-2-2-aminoacetamido-3-4-hydroxyphenyl propanoic acid, 2s-2-2-aminoacetyl amino-3-4-hydroxyphenyl propanoic acid
PubChem CID 92829
ChEBI CHEBI:73517
IUPAC Name (2S)-2-(2-aminoacetamido)-3-(4-hydroxyphenyl)propanoic acid
SMILES NCC(=O)N[C@@H](CC1=CC=C(O)C=C1)C(O)=O

Specifications

Color White to Yellow
Beilstein 2700715
Quantity 5 g
Formula Weight 238.24
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Glycyl-L-tyrosine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.