Learn More
N’-Nitrosopentyl-(3-picolyl)amine, TRC
Product Code. 30135850
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135850 25 mg 25mg
30009085 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135850 Supplier Toronto Research Chemicals Supplier No. TRCN54500025MG
 Restricted Item
Expédition estimée: 04-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 124521-15-9
Molecular Formula C11 H17 N3 O
Molecular Weight (g/mol) 207.27
Synonym N’-Nitrosopentyl-3-(Picolyl) Amine
IUPAC Name N-pentyl-N-(pyridin-3-ylmethyl)nitrous amide
SMILES CCCCCN(Cc1cccnc1)N=O
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C11H17N3O/c1-2-3-4-8-14(13-15)10-11-6-5-7-12-9-11/h5-7,9H,2-4,8,10H2,1H3
Formula Weight 207.1372
Chemical Name or Material N'-Nitrosopentyl-(3-picolyl)amine