Learn More
N1,N3-Bis(2,3-dihydroxypropyl)-5-(N-(2,3-dihydroxypropyl)acetamido)-2,4,6-triiodoisophthalamide, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 66108-95-0 |
| Molecular Formula | C19H26I3N3O9 |
| MDL Number | MFCD00077732 |
| Quantity | 250 mg |
| SMILES | O=C(C1=C(I)C(N(C(C)=O)CC(O)CO)=C(I)C(C(NCC(O)CO)=O)=C1I)NCC(O)CO |
| Molecular Weight (g/mol) | 821.14 |
| Percent Purity | 98% |
| Chemical Name or Material | N1,N3-Bis(2,3-dihydroxypropyl)-5-(N-(2,3-dihydroxypropyl)acetamido)-2,4,6-triiodoisophthalamide |