Learn More
Ohaus™ Navigator™
Product Code. 16263411 Shop All Ohaus Products
Change view
Click to view available options
Model: NVT22000-EU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16263411 Model: NVT22000-EU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 16263411 Supplier Ohaus™ Supplier No. 30456395
  • ONLINE EXCLUSIVE PRICE
    335.00€
    243.80€ / 1 pièces
    Économisez 91.20€
    27% de réduction
View all promotions
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
OHAUS™ Navigator™ is ideal for general weighing applications in laboratory, industrial, and education settings. This powerful balance can handle a diverse range of weighing applications with features such as fast stabilization time, overload protection at four times its rated capacity.
  • Navigator offers essential weighing applications at a competitive price point.
  • Advanced weighing technology offers one-second stabilization for fast and accurate weighing results, improving operator efficiency, productivity and throughput.
  • Overload protection system that can withstand loads up to four times the rated capacity safeguards the balance.
TRUSTED_SUSTAINABILITY
Specifications
Connection Type RS232, USB, or Ethernet (available as accessories)
Capacity 22,000 g
Display Type Backlit LCD
Readability 1.000000 g
Linearity 2.000000 g
Model NVT22000-EU
Pan Shape 230 x 174 mm
Repeatability 1.000000 g
Stabilization Time 1.000 s
Test Load 10 kg, OIML F2 or ASTM CL 4
Weighing Modes Carat, Grain, Gram, Kilogram, Newton, Ounce, Ounce Troy, Pennyweight, Pound, Pound:Ounce
Dimensions (L x W x H) 250 x 240 x 74 mm
Weight (Metric) 1.500 kg