Learn More
Weigert™ Neodisher™ S Defoamer
Product Code. 15158052
Change view
Click to view available options
Volume (Metric):
1 L
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15158052 1 L Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15158052 Supplier Weigert™ Supplier No. 430148
Quantity Price
1 397.80€ / 10 pièces
6 354.72€ / 10 pièces
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
To prevent foaming in alkaline cleaning solutions which are highly polluted by fat, protein, starch, sugar and others. Weigert™ Neodisher™ S Defoamer can be used in machine pre-cleaning as well as in main cleaning.
  • Based on emulsified silicone oil
  • For use in clinical and industrial laboratories, and the food and cosmetics industries
  • Avoid overdosing to prevent adhesion of silicone residues to surfaces
Specifications
Product Type Neodisher Antimousse
Volume (Metric) 1 L