Learn More
Nepafenac, TRC
Product Code. 30481660
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30481660 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30481660 Supplier Toronto Research Chemicals Supplier No. TRCN390080100MG
 Restricted Item
Expédition estimée: 09-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Nepafenac
CAS 78281-72-8
Molecular Formula C15 H14 N2 O2
Quantity 100 mg
Synonym Benzeneacetamide','2-amino-3-benzoyl-, 2-Amino-3-benzoylbenzeneacetamide, AHR 9434, AL 6515, Nepafenac, Nevanac
InChI Formula InChI=1S/C15H14N2O2/c16-13(18)9-11-7-4-8-12(14(11)17)15(19)10-5-2-1-3-6-10/h1-8H,9,17H2,(H2,16,18)
SMILES NC(=O)Cc1cccc(C(=O)c2ccccc2)c1N
IUPAC Name 2-(2-amino-3-benzoylphenyl)acetamide
Molecular Weight (g/mol) 254.28
Formula Weight 254.1055