Learn More
Nikethamide, MedChemExpress
Product Code. 16442329
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16442329 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16442329 Supplier Medchem Express Supplier No. HYB1280
 Restricted Item
Expédition estimée: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
MedChemExpress Nikethamide, one of the respiratory central stimulants, has the potential for respiratory failure research.
Nikethamide, one of the respiratory central stimulants, has the potential for respiratory failure research.
Specifications
Chemical Name or Material Nikethamide
CAS 59-26-7
Color Colorless
Purity Grade Notes Research
Molecular Formula C10H14N2O
Quantity 1 g
Synonym N,N-Diethylnicotinamide
Solubility Information H2O : ≥ 50 mg/mL (280.54 mM) ∣DMSO : ≥ 1 mg/mL (5.61 mM)
SMILES O=C(C1=CC=CN=C1)N(CC)CC
Molecular Weight (g/mol) 178.23
Formula Weight 178.23
Percent Purity 98.2%
Grade Research
Physical Form Liquid