Learn More
Nitisinone, MedChemExpress
Product Code. 16498548
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498548 10 mg 10mg
16444417 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16498548 Supplier Medchem Express Supplier No. HYB0607
 Restricted Item
Expédition estimée: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
MedChemExpress Nitisinone(SC0735) is an inhibitor of the enzyme 4-hydroxyphenylpyruvate dioxygenase.
Nitisinone(SC0735) is an inhibitor of the enzyme 4-hydroxyphenylpyruvate dioxygenase.
Specifications
Chemical Name or Material Nitisinone
Physical Form Powder
CAS 104206-65-7
Color Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C14H10F3NO5
For Use With (Application) Metabolism-sugar/lipid metabolism
Synonym NTBC Nitisone SC0735
Solubility Information DMSO : 41.67 mg/mL (126.57 mM; Need ultrasonic)
SMILES O=C1C(C(C2=CC=C(C(F)(F)F)C=C2[N+]([O-])=O)=O)C(CCC1)=O
Molecular Weight (g/mol) 329.23
Formula Weight 329.23
Percent Purity 98.76%
Grade Research