Learn More
Nitisinone, TRC
Product Code. 30006039
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30006039 100 mg 100mg
30009231 250 mg 250mg
30004350 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006039 Supplier Toronto Research Chemicals Supplier No. TRCN490135100MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 104206-65-7
Molecular Formula C14 H10 F3 N O5
Molecular Weight (g/mol) 329.23
Synonym 1,3-Cyclohexanedione, 2-[2-nitro-4-(trifluoromethyl)benzoyl]- (9CI, ACI), 2-[2-Nitro-4-(trifluoromethyl)benzoyl]-1,3-cyclohexanedione (ACI), Nitisinone, Nitisone, Nityr, NTBC, Orfadin, SC 0735
IUPAC Name 2-[2-nitro-4-(trifluoromethyl)benzoyl]cyclohexane-1,3-dione
SMILES [O-][N+](=O)c1cc(ccc1C(=O)C2C(=O)CCCC2=O)C(F)(F)F
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C14H10F3NO5/c15-14(16,17)7-4-5-8(9(6-7)18(22)23)13(21)12-10(19)2-1-3-11(12)20/h4-6,12H,1-3H2
Formula Weight 329.0511
Percent Purity >95%
Chemical Name or Material Nitisinone
Grade HPLC