Learn More
Lenz Laborglasintrumente Nitrogen Tubes (Schlenk Tubes) with NS Stopcock
Product Code. 10613442
Change view
Click to view available options
Standard Taper Joint:
NS 14/23 Cone
NS 14/23 Socket
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint Unit Size
10613442 NS 14/23 Socket Each
10376211 NS 14/23 Cone Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10613442 Supplier Lenz Laborglasintrumente Supplier No. 03370149
  • ONLINE EXCLUSIVE PRICE
    104.00€
    65.13€ / 1 pièces
    Économisez 38.87€
    37% de réduction
Expédition estimée: 14-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • With conical ground joint and NS stopcock
  • Made of DURAN™ tubing
Specifications
Capacity (Metric) 250 mL
Material Duran Glass
Standard Taper Joint NS 14/23 Socket