Learn More
Thermo Scientific™ [Nle13, Glu14] Motilin, Human, Porcine
Product Code. 15497559
Click to view available options
Quantity:
0.5 mg
1 mg
Unit Size:
0.5mg
1mg
This item is not returnable. View return policy
Product Code. 15497559
Brand: Thermo Scientific™ J66270.!D
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This product has been withdrawn. Where a replacement item has been identified it will be shown below
View alternative products
This item is not returnable. View return policy
For cellular and molecular biology applications
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Species Human, Porcine
Purification Method HPLC
Content And Storage -30°C to -10°C
Format Solid
Molecular Weight (g/mol) 2682.10
Product Type [Nle13, Glu14] Motilin, human, porcine
Quantity 0.5 mg
Regulatory Status RUO - research use only
Sensitivity Moisture sensitive
Sequence Phe-Val-Pro-Ile-Phe-Thr-Tyr-Gly-Glu-Leu-Gln-Arg-Nle-Glu-Glu-Lys-Glu-Arg-Asn-Lys-Gly-Gln
For Research Use Only