Learn More
Thermo Scientific™ Nonsterile Pipette Tips for Beckman™ Liquid Handling Systems
Product Code. 11751263 Shop All MBP Molecular BioProducts Products
Change view
Click to view available options
Volume (Metric):
20 μL
250 μL
For Use With (Equipment):
Beckman Coulter Biomek FX, Beckman Coulter Biomek NX SPAN-8
Fit for Purpose:
Certified Quality
Unit Size:
Pack of 4800
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) For Use With (Equipment) Fit for Purpose Unit Size
11751263 250 μL Beckman Coulter Biomek FX, Beckman Coulter Biomek NX SPAN-8 Certified Quality Pack of 4800
11711263 20 μL Beckman Coulter Biomek FX, Beckman Coulter Biomek NX SPAN-8 Certified Quality Pack of 4800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11751263 Supplier Thermo Scientific™ Supplier No. 919252
  • ONLINE EXCLUSIVE PRICE
    908.00€
    835.00€ / 4800 pièces
    Économisez 73.00€
    8% de réduction
View all promotions
Expédition estimée: 23-02-2027
Add to basket
Add to basket
Bulk Quote Request
This item is not returnable. View return policy
Increase pipetting performance through the superior tip straightness and low %CV of these pipette tips for automated liquid handling systems.
Increase pipetting performance with Thermo ScientificTM Nonsterile Pipette Tips for BeckmanTM Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. They provide accurate and precise performance because of superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.
Features:
  • Available in low retention and extended length
  • Lot-tested during production on actual workstation for which they are created
  • Extended length tips compatible with 96-, 384- and 1536- DeepWell™ plates
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 250 μL
Filtered Non-filtered
Sterility Non-sterile
Format Rack
Tip Style Conductive
For Use With (Equipment) Beckman Coulter Biomek FX, Beckman Coulter Biomek NX SPAN-8
Product Line Beckman
Fit for Purpose Certified Quality