Learn More
Thermo Scientific™ NP-40 lysis buffer with glycerol (2X)
Product Code. 15432969
Change view
Click to view available options
Quantity:
125 mL
250 mL
Unit Size:
125mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15432969 250 mL 250mL
15422969 125 mL 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15432969 Supplier Thermo Scientific™ Supplier No. J60143.AK
 Restricted Item
Expédition estimée: 21-12-2026
Add to basket
This item is not returnable. View return policy
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Color Colorless
Concentration 2X
Content And Storage Keep cold
Format Viscous Liquid
Quantity 250 mL