Learn More
Macherey-Nagel™ Nucleogel™ HPLC Column
Product Code. 10388251 Shop All Macherey-Nagel Products
Change view
Click to view available options
Unit Size:
Each
Material:
Nucleogel SAX 1000-8
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size Material
10388251 Each -
10286841 Each Nucleogel SAX 1000-8
10496281 Each -
10378631 Each -
10206511 Each -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10388251 Supplier Macherey-Nagel™ Supplier No. 719475
Expédition estimée: 16-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Base Material Polymer-based
Column Chemistry Polymer
For Use With (Application) Purification of Peptides, Large Proteins and Oligonucleotides, HPLC
Solvent Compatibility 0.1 M Na2SO4 + 0.2 % NaN3
Particle Size 8 μm
Stationary Phase Polymer-based
Type HPLC Column
Resin Composition Quaternized PEI