Learn More
Nudifloric Acid, TRC
Product Code. 30426779
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30426779 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30426779 Supplier Toronto Research Chemicals Supplier No. TRCN9254902.5G
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Nudifloric Acid
CAS 3719-45-7
Molecular Formula C7H7NO3
Quantity 2.5 g
Synonym 1,6-Dihydro-1-methyl-6-oxo-3-pyridinecarboxylic Acid, Methyl-6-oxo-1,6-dihydropyridine-3-carboxylic Acid, 1-Methyl-6-pyridone-3-carboxylic Acid, 1-Methylpyridin-2-one-5-carboxylic Acid
InChI Formula InChI=1S/C7H7NO3/c1-8-4-5(7(10)11)2-3-6(8)9/h2-4H,1H3,(H,10,11)
SMILES CN1C=C(C=CC1=O)C(=O)O
IUPAC Name 1-methyl-6-oxopyridine-3-carboxylic acid
Molecular Weight (g/mol) 153.14
Formula Weight 153.0426