Learn More
Thermo Scientific™ Nunc™ Disposable Loops and Needles, needle
BIOPHARMA
Product Code. 10769241
Change view
Click to view available options
Quantity:
Carton of 600
Unit Size:
Pack of 2400
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10769241 Carton of 600 Pack of 2400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10769241 Supplier Thermo Scientific™ Supplier No. 254399
  • ONLINE EXCLUSIVE PRICE
    497.00€
    418.99€ / 2400 pièces
    Économisez 78.01€
    15% de réduction
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Easily identify these color-coded disposable loops and needles, precision molded from nontoxic (USP Class VI) polystyrene material.
Easily identify color-coded Thermo Scientific™ Nunc™ Disposable Loops and Needles, precision molded from nontoxic (USP Class VI) polystyrene material to provide a smooth loop surface. Thermo Scientific Nunc products are high quality plastics for use in biotechnology, pharmaceutical and research laboratories, as well as in the production of vaccines and diagnostic kits.
Features:
  • Radiation sterilized—no need for flame sterilization
  • Hydrophilic treatment during manufacturing process
  • Liquid adherence properties that prevent accidental discharge of droplets
  • Loops available in 10 and 1μL sizes
  • Meet semi-quantitative standard for handling samples
  • Calibration documents available for any batch upon request
TRUSTED_SUSTAINABILITY
Specifications
Disposable Single-use
Material Polystyrene
Sterility Sterile
Color Yellow
Product Type Inoculating Needle
Quantity Carton of 600