Learn More
Thermo Scientific™ Nunc™ Screwdrivers for Cryobank and Bank-It™ Vial Systems, manual
BIOPHARMA
Product Code. 10709153
Change view
Click to view available options
Type:
Electric Screwdriver
Manual Screwdriver
Plug Type:
European
Not Applicable
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Plug Type Unit Size
10709153 Manual Screwdriver Not Applicable Each
10401633 Electric Screwdriver European Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10709153 Supplier Thermo Scientific™ Supplier No. 369643
  • ONLINE EXCLUSIVE PRICE
    60.05€
    47.34€ / 1 pièces
    Économisez 12.71€
    21% de réduction
View all promotions
 Restricted Item
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Quickly and easily cap and decap Thermo Scientific™ Nunc™ Cryobank tubes using these screwdrivers, available in manual and electric models to suit your application.
Quickly and easily cap and decap Nunc Cryobank tubes using Thermo Scientific™ Nunc™ Screwdrivers for Cryobank and Bank-It™ Systems, available in manual and electric models to suit your application. Nunc Cryobank vials provide the customer with superior tracking of their cryovials using a 2D code, the highest storage density on the market and the ability to automate the capping and decapping of the vials. The tubes click into the rack and can be handled without the risk of accidentally dropping a vial.
Screwdriver socket retains the vial cap, allowing one-handed manipulation of the cap.
TRUSTED_SUSTAINABILITY
Specifications
Description Manual Screwdriver
Coding 2D Barcode
Vial Type Cryobank, Bank-It
Color Red, Black
Plug Type Not Applicable
For Use With (Equipment) Cryobank and Bank-It Systems
Type Manual Screwdriver