Learn More
Zeiss™ Objective Plan
Product Code. 13469166
Change view
Click to view available options
Type:
Objective Plan S
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
13469166 Objective Plan S Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13469166 Supplier Zeiss™ Supplier No. 4352000000000
  • ONLINE EXCLUSIVE PRICE
    2055.00€
    1423.00€ / 1 pièces
    Économisez 632.00€
    30% de réduction
 Restricted Item
Expédition estimée: 29-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Benefit from the long experience of Zeiss in building world class optics to find the objectives dedicated for your application
Compatible Microscopes:
  • SteREO Discovery.V12
  • SteREO Discovery.V20
  • SteREO Discovery.V8
  • SteREO Lumar.V12
Specifications
Type Objective Plan S
Focus Distance 81 mm
For Use With (Equipment) Zeiss Microscopes
Magnification Power 1X
Quantity 1 Ea.