Learn More
Specac™ Omni-Cell™ CaF2 Liquid Cell Windows Rectangular
Product Code. 15269827
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Omni-Cell GS01800
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15269827 Omni-Cell GS01800 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15269827 Supplier Specac™ Supplier No. GS01812
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
This item is not returnable. View return policy
CaF2 rectangular Liquid Cell windows for use in the Omni-Cell GS01800. 41mm x 23mm and 4mm thick.
Specifications
For Use With (Equipment) Omni-Cell GS01800
Description 41 mm x 23 mm and 4 mm thick