Learn More
Thermo Scientific™ OMNIC™ Specta Software
Product Code. 18848251 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Analytical Raman
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
18848251 Analytical Raman Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18848251 Supplier Thermo Scientific™ Supplier No. 833036900
 Restricted Item
Expédition estimée: 29-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Quickly gather clear visual data as you work using the innovative identification and interpretation tools of the Thermo Scientific™ OMNIC™ Specta Software.
Quickly gather clear visual data as you work using the innovative identification and interpretation tools of Thermo Scientific™ OMNIC™ Specta Software.
OMNIC Specta Software creates a database of spectral data through is a unique combination of spectral identification tools, interpretation algorithms, and scientific documentation that indexes information on your hard disk. The software guides you in identifying materials and verifying assumptions—eliminating typical analytical tasks—so you get answers quickly and confidently.
The OMNIC Specta Software
  • Identify constituents in mixture samples with multi-component and contaminant searches — easier than tedious manipulations required by traditional search and subtract processing
  • Maintain historical data access to save time and prevent information loss 
  • Minimize time and effort with simple one click processing 
  • Receive immediate visual feedback for confidence in your results 
  • Ensure quality results with live displays for active data viewing 
  • Keep working during data collection for maximum productivity
  • Identify mixtures in one step for ultimate speed and confidence 
  • Communicate efficiently with modern PDF reports 
  • Copy directly into word processing programs
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Raman Spectrometers
Type Analytical Raman
For Use With (Application) Material Characterization