Learn More
- Home
- Products
- Chromatography Columns and Cartridges
- Low to Medium Pressure LC Columns
- Diba Omnifit™ EZ Chromatography Columns with Two Fixed Endpieces
Diba Omnifit™ EZ Chromatography Columns with Two Fixed Endpieces
Product Code. 15825344
Change view
Click to view available options
Length (Metric):
50 mm
100 mm
150 mm
250 mm
330 mm
400 mm
500 mm
Diameter (Metric) Inner:
6.6 mm
10 mm
15 mm
25 mm
35 mm
50 mm
Column Capacity:
0.7 mL
2.4 mL
4.1 mL
5.5 mL
7.5 mL
9.4 mL
10.3 mL
12.4 mL
12.7 mL
17.3 mL
21.2 mL
23.6 mL
29.1 mL
34.4 mL
36.9 mL
38.9 mL
53 mL
58.9 mL
65.4 mL
83.1 mL
108 mL
115.5 mL
147.3 mL
181.6 mL
211.7 mL
288.6 mL
356 mL
440.7 mL
600.9 mL
741.1 mL
941.4 mL
Unit Size:
Each
31 product options available for selection
| Product Code. | Length (Metric) | Diameter (Metric) Inner | Column Capacity | Unit Size |
|---|---|---|---|---|
| 15825344 | 100 mm | 10 mm | 5.5 mL | Each |
| 15855344 | 150 mm | 10 mm | 9.4 mL | Each |
| 15815384 | 150 mm | 25 mm | 58.9 mL | Each |
| 15865394 | 150 mm | 35 mm | 115.5 mL | Each |
| 15855404 | 400 mm | 35 mm | 356 mL | Each |
| 15845354 | 400 mm | 10 mm | 29.1 mL | Each |
| 15815414 | 330 mm | 50 mm | 600.9 mL | Each |
| 15875354 | 500 mm | 10 mm | 36.9 mL | Each |
| 15845414 | 400 mm | 50 mm | 741.1 mL | Each |
| 15895394 | 250 mm | 35 mm | 211.7 mL | Each |
| 15895334 | 400 mm | 6.6 mm | 12.7 mL | Each |
| 15855374 | 500 mm | 15 mm | 83.1 mL | Each |
| 15845384 | 250 mm | 25 mm | 108 mL | Each |
| 15885374 | 100 mm | 25 mm | 34.4 mL | Each |
| 15825404 | 330 mm | 35 mm | 288.6 mL | Each |
| 15805334 | 150 mm | 6.6 mm | 4.1 mL | Each |
| 15875324 | 100 mm | 6.6 mm | 2.4 mL | Each |
| 15825374 | 400 mm | 15 mm | 65.4 mL | Each |
| 15805364 | 100 mm | 15 mm | 12.4 mL | Each |
| 15835334 | 250 mm | 6.6 mm | 7.5 mL | Each |
| 15885404 | 250 mm | 50 mm | 440.7 mL | Each |
| 15815354 | 330 mm | 10 mm | 23.6 mL | Each |
| 15895364 | 330 mm | 15 mm | 53 mL | Each |
| 15805394 | 400 mm | 25 mm | 181.6 mL | Each |
| 15865364 | 250 mm | 15 mm | 38.9 mL | Each |
| 15845324 | 50 mm | 6.6 mm | 0.7 mL | Each |
| 15835364 | 150 mm | 15 mm | 21.2 mL | Each |
| 15865334 | 330 mm | 6.6 mm | 10.3 mL | Each |
| 15885344 | 250 mm | 10 mm | 17.3 mL | Each |
| 15875414 | 500 mm | 50 mm | 941.4 mL | Each |
| 15875384 | 330 mm | 25 mm | 147.3 mL | Each |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 31 options available.
This item is not returnable.
View return policy
Product Code.
15825344
Supplier
Diba
Supplier No.
006EZ1010FF
Expédition estimée: 18-12-2026
Connectez-vous pour voir le stock disponible
Connectez-vous pour voir le stock disponible
Add to basket
Add to basket
This item is not returnable.
View return policy
Description
- Precision borosilicate glass columns provide inertness, visibility, and performance
- Superior frit placement adds bed support and even distribution of sample flow
- Reusable columns let you pack your own media specific to your application
Specifications
Specifications
| Max. Pressure | 600 psi |
| Pore Size | 10 and 30 μm |
| Column Capacity | 5.5 mL |
| Length (Metric) | 100 mm |
| Material | Borosilicate Glass, PTFE, Polyethylene, FKM/FPM |
| Max. Temperature | 40°C |
| Includes | Two (0.5 m and 2 m L) 1/16 in. OD PTFE tubing assembly with 1/4-28 UNF Polypropylene fitting at one end, a frit kit (contains two each of 10 μm PE, 10 μm PTFE, and 30 μm PTFE frits), and a Click-N-Seal™ 10-32 PEEK conical port fitting. |
| Temperature (Metric) Range | 4 to 40°C |
| Type | Liquid Chromatography Column |
| Diameter (Metric) Inner | 10 mm |