Learn More
Thermo Scientific™ Orion™ Pro Star Series Meter Accessories
Product Code. 30245782
Change view
Click to view available options
Type:
Electrode Stand/Holder
Holder Head
Power Supply
Rugged Protective Display Cover
Standard Protective Display Cover
USB Barcode Scanner
USB Computer Cable
USB Flash Drive
USB Keyboard
USB Mouse
Weighted Base
Unit Size:
1 set
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
30245782 Electrode Stand/Holder 1 set
30245820 Rugged Protective Display Cover 1 set
30245792 Standard Protective Display Cover 1 set
30245797 Power Supply 1 set
30245788 Holder Head 1 set
30245821 Weighted Base 1 set
30245783 USB Barcode Scanner 1 set
30245781 USB Flash Drive 1 set
30245796 USB Keyboard 1 set
30245784 USB Mouse 1 set
30245777 USB Computer Cable 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 30245782 Supplier Thermo Scientific™ Supplier No. PSTARARM
  • ONLINE EXCLUSIVE PRICE
    183.00€
    151.20€ / 1 kits
    Économisez 31.80€
    17% de réduction
 Restricted Item
En stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Select Thermo Scientific™ Orion™ Pro Star series meter accessories to support your Orion Pro Star PH211, EC212, DO213, PI214, PC215, PD216, PI217 and MP218 bench meters.
Select Orion Pro Star series meter accessories to support your Orion Pro Star PH211, EC212, DO213, PI214, PC215, PD216, PI217 and MP218 bench meters. Add an optional accessory to input sample ID with the USB barcode scanner, USB keyboard or USB mouse. Replace included meter accessories such as the universal power adapter, USB flash drive, USB computer cable and electrode stand with base. Select a standard or rugged protective display cover to add additional protection for your meter in challenging lab conditions.
Select Orion Pro Star series meter accessories to support your Orion Pro Star bench meters. All listed accessories are compatible with the following meter models:
  • Orion Pro Star PH211 pH/ORP bench meter
  • Orion Pro Star EC212 conductivity bench meter
  • Orion Pro Star DO213 dissolved oxygen bench meter
  • Orion Pro Star PI214 pH/ion bench meter
  • Orion Pro Star PC215 pH/ion and conductivity bench meter
  • Orion Pro Star PD216 pH/ion and DO bench meter
  • Orion Pro Star PI217 dual channel pH/ion bench meter
  • Orion Pro Star MP218 pH/ion, conductivity and DO bench meter
Orion Pro Star Series Meter Optional Accessories
  • PSTAR-UBS Orion Pro Star series USB barcode scanner is tested to be compatible with Orion Pro Star meters and allows sample ID values to be easily entered into the meter from barcoded samples.
  • PSTAR-UFD Orion Pro Star series USB flash drive is tested to be compatible with Orion Pro Star meters and allows data to be transferred from the meter to a computer, methods to be transferred between meters and software updates to be transferred from the web to the meter.
  • PSTAR-UKB Orion Pro Star series USB keyboard is tested to be compatible with Orion Pro Star meters and allows various numeric and alphanumeric values including sample ID values to be easily entered into the meter.
  • PSTAR-UMS Orion Pro Star series USB mouse is tested to be compatible with Orion Pro Star meters and allows the meter to be operated with a remote device, removing the need to touch the display to operate the meter, which can be helpful in environments where sample carryover may damage the meter.
  • PSTAR-PCR Orion Pro Star series rugged protective display cover is designed to add additional protect to the meter display during operation, which can be helpful in environments where the display may become dirty or scratched in the lab.
Orion Pro Star Series Meter Replacement Accessories
  • PSTAR-ARM Orion Pro Star series electrode stand with base allows you to replace your complete electrode stand as needed.
  • PSTAR-SHH Orion Pro Star series holder head allows you to replace just the holder head component of the electrode stand as needed.
  • PSTAR-SWB Orion Pro Star series weighted base allows you to replace just the base of the electrode stand as needed.
  • PSTAR-PCS Orion Pro Star series standard protective display cover allows you to replace you display cover as needed.
  • PSTAR-PWR Orion Pro Star series universal power supply allows you to replace your power supply as needed.
  • PSTAR-USB Orion Pro Star series USB computer cable allows you to replace your meter-to-computer cable as needed.
Additional Accessories Compatible with Orion Pro Star Series Meters
  • 096019-WA Orion stirrer probe for bench meters
  • STARA-106 Orion compact ink printer with meter-to-printer cable, ink, paper and 100-240V 50/60 Hz power supply
TRUSTED_SUSTAINABILITY
Specifications
Type Electrode Stand/Holder
For Use With (Application) Replace your complete electrode stand as needed
Includes Weighted base, electrode arm with cable management system, electrode holder head with clip
Compatibility pH electrodes, ORP electrodes, ion selective electrodes (ISE) and other electrodes with a 12mm electrode body diameter; Orion ATC temperature probe; Orion stirrer probe; conductivity electrodes, polarographic dissolved oxygen electrodes and other electrodes with a 15mm diameter; gas-sensing ion selective electrodes that require a 20° angle holder; larger electrodes including the optical dissolved oxygen electrode
For Use With (Equipment) Orion Pro Star series bench electrochemistry meters