Learn More
Thermo Scientific™ Orion™ Star A Series Benchtop Meter-Attached Electrode Stand
Product Code. 11641489
Change view
Click to view available options
Electrode Arm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11641489 Electrode Arm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11641489 Supplier Thermo Scientific™ Supplier No. STARABEA
  • ONLINE EXCLUSIVE PRICE
    175.00€
    165.00€ / 1 pièces
    Économisez 10.00€
    5% de réduction
View all promotions
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Use the Thermo Scientific™ Orion™ Star A Series Benchtop Meter-Attached Electrode Stand to easily place sensors in samples, simplify rinsing and storage and minimize breakage
Use the Thermo Scientific™ Orion™ Star A Series Benchtop Meter-Attached Electrode Stand to easily place sensors in samples, simplify rinsing and storage between samples and minimize electrode breakage using the stand's cable management system. Attach the stand to either side of Thermo Scientific™ Orion™ Versa Star benchtop meters and Thermo Scientific™ Orion™ Star A210 and Star A110 series benchtop meters or use the stand with weighted base (Cat. No. STARA-HB) for compatibility with any meter system. Includes meter bracket, arm and electrode holder (Cat. No. STARA-ESCD).
TRUSTED_SUSTAINABILITY
Specifications
Description Orion Star A series meter stand, electrode holder and meter-attachable base
Type Electrode Stand/Holder
Includes Redesigned electrode holder (Cat .No. STARA-ELHD), plastic stand with cable management system and meter-attachable base
Recommended Accessories To use with almost any Orion benchtop or portable meter, purchase the weighted base separately (Cat. No. STARA-HB)
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.